EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
The ADMA/DDAH/NO pathway in human vein endothelial cells exposed to arsenite.
PMID 28502835
Abstract
Inorganic arsenic (iAs) exposure is related to cardiovascular disease, which is characterized by endothelial dysfunction and nitric oxide (NO) depletion. The mechanisms underlying NO depletion as related to iAs exposure are not fully understood. The endogenous inhibitor of nitric oxide synthase, asymmetric dimethylarginine (ADMA), might be a molecular target of iAs. ADMA concentrations are regulated by proteins involved in its synthesis (arginine methyl transferase 1 [PRMT-1]) and degradation (dimethylarginine dimethylaminohydrolase [DDAH]). Both, ADMA and NO are susceptible to oxidative stress. We aimed to determine the ADMA/DDAH/NO pathway in human vein endothelial cells (HUVEC-CS) exposed to arsenite. We exposed HUVEC-CS cells to 1, 2.5 and 5μM of arsenite for 24h. We proved that arsenite at 5μM was able to decrease NO levels with an associated increase in ADMA and depletion of l-arginine in HUVEC-CS cells. We also found a decrease in DDAH-1 protein expression with 5μM of arsenite compared to the control group. However, we did not observe significant differences in PRMT-1 protein expression at any of the concentrations of arsenite employed. Finally, arsenite (2.5 and 5μM) increased NADPH oxidase 4 protein levels compared with the control group. We conclude that ADMA, l-arginine and DDAH are involved in NO depletion produced by arsenite, and that the mechanism is related to oxidative stress.