EMAIL THIS PAGE TO A FRIEND
Journal of Muscle Research and Cell Motility
Okadaic acid. Protein phosphatase inhibition and muscle contractile effects.
PMID 2850299
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19-130 Okadaic Acid (sodium salt)