EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
Pioppino mushroom in southern Italy: an undervalued source of nutrients and bioactive compounds.
PMID 28503801
Abstract
Agrocybe aegerita (V. Brig.) Singer, commonly known as Pioppino, is a popular edible mushroom, known in the Campania Region (Italy). Despite its habitual consumption, little nutritional and biochemical information is available. Thus, nutritional values, anti-radical properties and chemical composition of the wild Pioppino were compared to those of the cultivated Agaricus bisporus (J.E. Lange) Imbach (known as Champignon), equally analysed. Macronutrient components (proteins, carbohydrates and lipids), free and protein amino acids and fatty acid content of poplar mushroom were achieved. Total phenol content of a defatted Pioppino alcoholic extract (PM) was determined, whereas DPPH and ABTS methods were applied to determine the radical scavenging capabilities of the extract. Ferricyanide and ORAC-fluorescein methods were also performed. Finally, LC-HRMS was used to identify and quantify the main metabolites in the extract. PM was mainly constituted of disaccharides, hexitol derivatives and malic acid. Coumaric acid isomers and C All data revealed that wild Pioppino is an excellent functional food, by far exceeding that of the Champignon. Therefore, these data are useful to promote the consumption of this mushroom encouraging thus its biological cultivation, due to wild availability is strongly compromised by the extensive use of fungicides. © 2017 Society of Chemical Industry.