EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Pyrazolo[4,3-h]quinoline Ligand-Based Iridium(III) Complexes for Electrochemiluminescence.
PMID 28503856
Abstract
Electrochemiluminescence (ECL) based on [Ru(bpy)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F408
Ferrocene, 98%
C10H10Fe