EMAIL THIS PAGE TO A FRIEND
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes
[Ferrocene, ruthenocene and rhodocene analogs in haloperidol synthesis and organ distribution after labeling with 103Ru and 103mRh].
PMID 2851003
Abstract
Ferrocene-Haloperidol was synthesized by N-alkylation of 4-(4'-chlorophenyl)- 4-hydroxypiperidine with 1-ferrocenyl-4-chlor-butan-1-on. By heating the ferrocene-haloperidol with 103RuCl3 the 103Ru-labelled ruthenocene-haloperidol was obtained. This compound showed a high affinity for lung but not for brain in rats and mice. The decay of the 103Ru labelled compound results in the formation of the 103mRh labelled rhodocene-haloperidol, which is rapidly oxidized by air to the corresponding rhodocinium-haloperidol. This compound can be separated by extraction and TLC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262455
Bis(cyclopentadienyl)ruthenium(II), 97%
C10H10Ru