EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Agrochemical loading in drains and rivers and its connection with pollution in coastal lagoons of the Mexican Pacific.
PMID 28510105
Abstract
The state of Sinaloa in Mexico is an industrialized agricultural region with a documented pesticide usage of 700 t year
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45403
Coumaphos, PESTANAL®, analytical standard
C14H16ClO5PS