Botanical studies
Chilling susceptibility in mungbean varieties is associated with their differentially expressed genes.
PMID 28510190
Mungbean (Vigna radiata L. Wilczek) is an economically important legume of high nutritional value, however, its cultivation is limited by susceptibility to chilling. Varieties NM94 and VC1973A, with differential susceptibility to stress, serve as good materials for uncovering how they differ in chilling tolerance. This study aimed to identify the ultrastructural, physiological and molecular changes to provide new insights on the differential susceptibility to chilling between varieties VC1973A and NM94. Chilling stress caused a greater reduction in relative growth rate, a more significant decrease in maximum photochemical efficiency of PSII and DPPH scavenging activity and more-pronounced ultrastructural changes in VC1973A than in NM94 seedlings. Comparative analyses of transcriptional profiles in NM94 and VC1973A revealed that the higher expression of chilling regulated genes (CORs) in NM94. The transcript levels of lipid transfer protein (LTP), dehydrin (DHN) and plant defensin (PDF) in NM94 seedlings after 72xa0h at 4xa0°C was higher than that in its parental lines VC1973A, 6601 and VC2768A. Our results suggested that LTP, DHN and PDF may mediate chilling tolerance in NM94 seedlings.