EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Inactivation of the KSRP gene modifies collagen antibody induced arthritis.
PMID 28511090
Abstract
The KH type splicing regulatory protein (KSRP) is a nucleic acid binding protein, which negatively regulates the stability and/or translatability of many mRNA species encoding immune-relevant proteins. As KSRP is expressed in immune cells including T and B cells, neutrophils, macrophages and dendritic cells, we wanted to analyze its importance for the development of autoimmune diseases. We chose collagen antibody-induced arthritis (CAIA) as an appropriate autoimmune disease mouse model in which neutrophils and macrophages constitute the main effector cell populations. We compared arthritis induction in wild type (WT) and KSRP