EMAIL THIS PAGE TO A FRIEND
Food & function
l-Ornithine stimulates growth hormone release in a manner dependent on the ghrelin system.
PMID 28513740
Abstract
We found that intraduodenal administration of l-ornithine (l-Orn) stimulates growth hormone (GH) secretion in Wistar rats, and then investigated its mechanism. GH-releasing activity after intraduodenal administration of l-Orn was blocked by [d-Lys
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8688
(S)-(−)-Propranolol hydrochloride, ≥98% (TLC), powder
C16H21NO2 · HCl