EMAIL THIS PAGE TO A FRIEND
Food & function
Re-assembled casein micelles improve in vitro bioavailability of vitamin D in a Caco-2 cell model.
PMID 28513755
Abstract
The pandemic of vitamin D (VD) deficiency, and the global rise in obesity stimulate a need for staple low-fat foods and beverages enriched with VD. In light of consumer demand for a clean label, the use of natural endogenous food ingredients as delivery vehicles is of great interest. To this end, re-assembled casein micelles (rCM) have been shown to help retain VD during processing and shelf life and provide high bioavailability in low-fat milk and non-fat yoghurt. This follow-up study focused on the performance of VD-loaded rCM after drying and reconstitution, considering VD retention during simulated digestion, and the subsequent in vitro bioavailability of the vitamin. rCM conferred great protection to VD