EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Progerin sequestration of PCNA promotes replication fork collapse and mislocalization of XPA in laminopathy-related progeroid syndromes.
PMID 28515154
Abstract
Hutchinson-Gilford progeria syndrome (HGPS) is a rare genetic disorder that is caused by a point mutation in the
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N4411
NU6027, ≥98% (HPLC)
C11H17N5O2