EMAIL THIS PAGE TO A FRIEND
Journal of applied microbiology
Differences between two strains of Ceriporiopsis subvermispora on improving the nutritive value of wheat straw for ruminants.
PMID 28517113
Abstract
This study evaluated differences between two strains of Ceriporiopsis subvermispora on improving the nutritive value and in vitro degradability of wheat straw. Wheat straw was treated with the fungi for 7 weeks. Weekly samples were analysed for ergosterol content, in vitro gas production (IVGP), chemical composition and lignin-degrading enzyme activity. Ergosterol data showed CS1 to have a faster initial growth than CS2 and reaching a stationary phase after 3 weeks. The IVGP of CS1-treated wheat straw exceeded the control earlier than CS2 (4 vs 5 weeks). CS1 showed a significantly higher (P < 0·001) selectivity in lignin degradation compared to CS2. Both strains showed peak activity of laccase and manganese peroxidase (MnP) at week 1. CS1 showed a significantly higher (P < 0·001) laccase activity, but lower (P = 0·008) MnP activity compared to CS2. Both CS strains improved the nutritive value of wheat straw. Variation between strains was clearly demonstrated by their growth pattern and enzyme activities. The differences among the two strains provide an opportunity for future selection and breeding programs in improving the extent and selectivity of lignin degradation in agricultural biomass.