EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Increased in vivo synthesis of retinal Na pump isoforms after pre-treatment with citrate buffer.
PMID 2851793
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30030
Cymarin, ≥96% (HPLC)
C30H44O9