EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Antimicrobial agent-free hybrid cationic starch/sodium alginate polyelectrolyte films for food packaging materials.
PMID 28521996
Abstract
This study aimed to develop polyelectrolyte structured antimicrobial food packaging materials that do not contain any antimicrobial agents. Cationic starch was synthesized and characterized by FT-IR spectroscopy and
Related Materials