Cancer research
Targeting FBW7 as a Strategy to Overcome Resistance to Targeted Therapy in Non-Small Cell Lung Cancer.
PMID 28522751
Inhibition of EGFR and anaplastic lymphoma kinase (ALK) signaling is highly effective in a subgroup of non-small cell lung cancer (NSCLC) patients with distinct clinicopathologic features. However, resistance to EGFR and ALK inhibitors inevitably occurs, and the molecular mechanism underlying resistance is not fully understood. In this study, we report a PI3K/Akt- and MEK/ERK-independent resistance mechanism by which loss of the E3 ubiquitin ligase F-box and WD repeat domain containing 7 (FBW7α) leads to targeted therapy resistance via stabilization of antiapoptotic protein MCL-1. Using a panel of in vitro and in vivo studies, we showed that the regulatory machinery responsible for MCL-1 protein degradation was a step-wise event involving phosphorylation and nucleus translocation. ERK cooperated with GSKβ to phosphorylate MCL-1 Ser159 residue, which enabled MCL-1 to translocate into the nucleus and bind FBW7. Defects in this sequence impaired MCL-1 degradation and cell apoptosis, recapitulating phenotypes observed in FBW7 deficiency. Downregulation of FBW7 was found in EGFR inhibitor-resistant human NSCLC specimens and correlated with increased MCL-1 protein expression. Reactivation of FBW7 sensitized resistant cells to targeted therapy and facilitated MCL-1 degradation. Overall, our study provides proof-of-principle insight into a PI3K/Akt- and MEK/ERK-independent resistant model and suggests that targeting FBW7 can overcome resistance to targeted therapy. Cancer Res; 77(13); 3527-39. ©2017 AACR.