EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Novel amidino substituted benzimidazole and benzothiazole benzo[b]thieno-2-carboxamides exert strong antiproliferative and DNA binding properties.
PMID 28525845
Abstract
Within this manuscript design, synthesis of novel 2-imidazolinyl substituted benzo[b]thieno-2-carboxamides bearing either benzimidazole or benzothiazole subunit and biological activity are presented and described. The antiproliferative activities were assessed in vitro on a panel of human cancer cell lines. Tested compounds showed moderate activity while cytotoxicity on normal fibroblasts was lower in comparison with 5-fluorouracile. The variations of 2-imidazolinyl substituent at heteroaromatic subunits in different positions led to different cytotoxic properties. The strongest selective activity against HeLa cells was observed for the benzothiazole derivative 4d with 2-imidazolinyl group at the benzo[b]thiophene subunit with a corresponding IC
Related Materials