EMAIL THIS PAGE TO A FRIEND
Toxicon : official journal of the International Society on Toxinology
Activation of Na
PMID 28526336
Abstract
We previously reported unexpected apoptosis-like cell death induced by nematocyst venom (NV) from Chrysaora helvola Brandt (C. helvola) jellyfish. To assess whether the pore formation mechanism underlay the action of NV, the change in cell membrane permeability was studied in both chicken erythrocytes and human CNE-2 cells. Initially, paradoxical results were derived from osmoprotectant protection assays. Polyethylene glycol (PEG)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
FD40 Fluorescein isothiocyanate–dextran, average mol wt 40,000