EMAIL THIS PAGE TO A FRIEND
Journal of vascular surgery
A single nucleotide polymorphism of cyclin-dependent kinase inhibitor 1B (p27
PMID 28526559
Abstract
Cyclin-dependent kinase inhibitor 1B (p27 Human saphenous vein adventitial cells and intimal/medial smooth muscle cells (SMCs) were derived from explants obtained at the time of lower extremity bypass operations. We determined the following in adventitial cells and SMCs as a function of the p27 SNP genotype: (1) p27 promoter activity, (2) p27 messenger (m)RNA and protein levels, and (3) growth and collagen gel contraction. Deoxyribonuclease I footprinting was also performed in adventitial cells and SMCs. p27 promoter activity, deoxyribonuclease I footprinting, p27 mRNA levels, and p27 protein levels demonstrated that the p27 SNP is functional in adventitial cells and SMCs. Both cell types with the graft failure protective AA genotype had more p27 mRNA and protein. As predicted because of higher levels of p27 protein, adventitial cells with the AA genotype grew slower than those of the CC genotype. Unexpectedly, SMCs did not show this genotype-dependent growth response. These results support the functionality of the p27 SNP in venous SMCs and adventitial cells, but an effect of the SNP on cell proliferation is limited to only adventitial cells. These data point to a potential role for adventitial cells in human vein graft failure and also suggest that SMCs express factors that interfere with the activity of p27.