EMAIL THIS PAGE TO A FRIEND
Plant physiology and biochemistry : PPB
Monochromatic green light induces an aberrant accumulation of geranylgeranyled chlorophylls in plants.
PMID 28527413
Abstract
Light quality is an important environmental factor affecting the biosynthesis of photosynthetic pigments whose production seems to be affected not only quantitatively but also qualitatively. In this work, we set out to identify unusual pigment detected in leaves of barley (Hordeum vulgare L.) and explain its presence in plants grown under monochromatic green light (GL; 500-590 nm). The chromatographic analysis (HPLC-DAD) revealed that a peak belonging to this unknown pigment is eluted between chlorophyll (Chl) a and b. This pigment exhibited the same absorption spectrum and fluorescence excitation and emission spectra as Chl a. It was negligible in control plants cultivated under white light of the same irradiance (photosynthetic photon flux density of 240 μmol m