EMAIL THIS PAGE TO A FRIEND
IET nanobiotechnology
Green synthesis of silver nanoparticles using bovine skin gelatin and its antibacterial effect on clinical bacterial isolates.
PMID 28530191
Abstract
Nanoparticles are being increasingly used in day-to-day life. Therefore, concerns have been raised regarding their interactions with the surrounding environment. This study focused on a simple green method for synthesizing silver nanoparticles (Ag-NPs) in an autoclave at 15 psi (103 kPa) and 121°C. An aqueous solution of AgNO