EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Immunomodulatory function of whole human umbilical cord derived mesenchymal stem cells.
PMID 28531813
Abstract
Bone marrow derived mesenchymal stem cells (MSCs) play a critical role in immune modulation. However, immunomodulatory function of whole human umbilical cord derived mesenchymal stem cells (UC-MSCs) remains unclear. In this study, UC-MSCs were separated from whole umbilical cord using a single enzyme digestion. UC-MSCs (CD73