EMAIL THIS PAGE TO A FRIEND
Materials Science & Engineering. C, Materials For Biological Applications
Electrochemically reduced graphene oxide/Poly-Glycine composite modified electrode for sensitive determination of l-dopa.
PMID 28532045