EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Serum amyloid A: an ozone-induced circulating factor with potentially important functions in the lung-brain axis.
PMID 28533327
Abstract
Accumulating evidence suggests that O
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G10
Spectrophotometer cuvettes, glass, Without stopper