EMAIL THIS PAGE TO A FRIEND
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging
Repeatability of [
PMID 28534214
Abstract
We studied the effect of varying specific activity of [ Athymic nude mice bearing PC3-PIP (PSMA+) and PC3 (PSMA-) bilateral flank tumors were assessed to study intra- and inter-day repeatability of [ Region of interest (ROI) analysis of the [ This study demonstrates that [