EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Modulation of the Conformational Dynamics of Apo-Adenylate Kinase through a π-Cation Interaction.
PMID 28534408
Abstract
Large-scale conformational transition from open to closed state of adenylate kinase (ADK) is essential for its catalytic cycle. Apo-ADK undergoes conformational transition in a way that closely resembles an open-to-closed conformational transition. Here, equilibrium simulations, free-energy simulations, and quantum mechanics/molecular mechanics (QM/MM) calculations in combination with several bioinformatics approaches have been used to explore the molecular origin of this conformational transition in apo-ADK. In addition to its conventional open state, Escherichia coli apo-ADK adopts conformations that resemble a closed-like intermediate, the "half-open-half-closed" (HOHC) state, and a π-cation interaction can account for the stability of this HOHC state. Energetics and the electronic properties of this π-cation interaction have been explored using QM/MM calculations. Upon rescinding the π-cation interaction, the conformational landscape of the apo-ADK changes completely. The apo-ADK population is shifted completely toward the open state. This π-cation interaction is highly conserved in bacterial ADK; the cationic guanidinium moiety of a conserved ARG interacts with the delocalized π-electron cloud of either PHE or TYR. Interestingly, this study demonstrates the modulation of a principal protein dynamics by a conserved specific π-cation interaction across different organisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69480
N-Methyl-N-trimethylsilylacetamide, for GC derivatization, ≥97.0% (GC)
C6H15NOSi