EMAIL THIS PAGE TO A FRIEND
Oncology reports
MicroRNA-200c regulates cisplatin resistance by targeting ZEB2 in human gastric cancer cells.
PMID 28534959
Abstract
This study was specifically designed to confirm the hypothesis that microRNA-200c (miR-200c) affects the development of cisplatin (DDP) resistance in human gastric cancer cells by targeting zinc finger E-box binding homeobox 2 (ZEB2). A total of 50 gastric cancer tissues and their corresponding normal adjacent tissue samples were collected. Then, the expression levels of miR-200c and ZEB2 in both gastric cancer specimens and cells were detected using the quantitative real-time reverse transcription-polymerase chain reaction (qRT-PCR) and immunohistochemical methods. A dual‑luciferase reporter gene assay was conducted to evaluate the effect of miR-200c on the 3'-untranslated region (3'UTR) luciferase activity of ZEB2. SGC7901/DDP cells were transfected with miR-200c mimics and ZEB2 siRNA, respectively. Subsequently, changes in cellular proliferation and apoptosis were detected through the methyl thiazolyl tetrazolium assay and flow cytometric analysis, respectively. We also carried out a western blot analysis assay in order to detect the expression of apoptosis-related genes and ZEB2. miR-200c was significantly downregulated and ZEB2 was significantly upregulated in both gastric cancer tissues and SGC7901/DDP cells when compared with those in normal tissues and SGC7901 cells (P<0.01). The dual luciferase reporter gene assay showed that miR-200c could specifically bind with the 3'UTR of ZEB2 and significantly suppress the luciferase activity by 42% (P<0.01). Upregulation of miR-200c or downregulation of ZEB2 enhanced the sensitivity of SGC7901/DDP cells to DDP. miR‑200c was significantly downregulated in both gastric cancer tissues and cells, while the expression of ZEB2 exhibited the opposite trend. Our study further demonstrated that miR-200c could enhance the sensitivity of SGC7901/DDP cells to DDP through targeted regulation of ZEB2 expression in gastric cancer tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU010131 MISSION® esiRNA, esiRNA targeting human ZEB2 (esiRNA1)