EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Inhibitory Effect of Piceatannol on TNF-α-Mediated Inflammation and Insulin Resistance in 3T3-L1 Adipocytes.
PMID 28535046
Abstract
Piceatannol, a bioactive component in grape and blueberry, was examined for its potential in decreasing the inflammatory activities in adipocytes using a cocultured adipocyte and macrophage system, and suppressing tumor necrosis factor-α (TNF-α)-mediated inflammation and the related insulin resistance using a 3T3-L1 adipocyte model. Piceatannol at 10 μM significantly reduced the release of inflammatory cytokines of TNF-α and monocyte chemoattractant protein-1 (MCP-1) by 19 and 31% in the cocultured system, respectively. Pretreatment with piceatannol also inhibited TNF-α-induced expression of interleukin-6 (IL-6) and MCP-1 at both mRNA and protein levels in the 3T3-L1 adipocytes. Piceatannol also partially improved the malfunction of insulin-stimulated glucose uptake, which was reduced by TNF-α in 3T3-L1 adipocytes. Furthermore, the inhibitions were mediated by significant blocking of IκBα phosphorylation and nuclear factor-κB (NF-κB) activation through suppressing nuclear translocation of NF-κB p65 along with c-Jun N-terminal kinase (JNK)-mitogen activated protein kinase (MAPK) activation. In addition, the Akt-dependent forkhead box O1 (FoxO1) signaling pathway was involved in the restoration of insulin-stimulated glucose uptake through suppressing the down-regulation of phosphorylation of Akt and FoxO1 expressions. These results suggested the potential of piceatannol in improving chronic inflammatory condition and insulin sensitivity in obese adipose tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
861677
DL-Cysteine, technical grade
C3H7NO2S
P0453
Piceatannol, powder
C14H12O4