Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
MiR-129-5p Inhibits Proliferation and Invasion of Chondrosarcoma Cells by Regulating SOX4/Wnt/β-Catenin Signaling Pathway.
PMID 28535514
Recently, microRNAs (miRNA) have been identified as novel regulators in Chondrosarcoma (CHS). This study was aimed to identify the roles of miR-129-5p-5p in regulation of SOX4 and Wnt/β-catenin signaling pathway, as well as cell proliferation and apoptosis in chondrosarcomas. Tissue samples were obtained from chondrosarcoma patients. Immunohistochemistry, real-time quantitative RT-PCR (RT-qPCR) and western blot analysis were performed to detect the expressions of miR-129-5p and SOX4. Luciferase assay was conducted to confirm that miR-129-5p directly targeted SOX4 mRNA. Manipulations of miR-129-5p and SOX4 expression were achieved through cell transfection. Cell proliferation, migration and apoptosis were evaluated by CCK-8 assay, colony forming assay, wound healing assay and flow cytometry in vitro. For in vivo experiment, the tumor xenograft model was established to evaluate the effects of miR-129-5p and SOX4 on chondrosarcomas. The expression of miR-129-5p was significantly down-regulated in chondrosarcoma tissues as well as cells in comparison with normal ones, while SOX4 was over-activated. Further studies suggested that miR-129-5p suppressed cell proliferation, migration and promoted apoptosis by inhibiting SOX4 and Wnt/β-catenin pathway. MiR-129-5p inhibits the Wnt/β-catenin signaling pathway by targeting SOX4 and further suppresses cell proliferation, migration and promotes apoptosis in chondrosarcomas.