EMAIL THIS PAGE TO A FRIEND
Nanoscale research letters
Realizing Full Coverage of Stable Perovskite Film by Modified Anti-Solvent Process.
PMID 28535603
Abstract
Lead-free solution-processed solid-state photovoltaic devices based on formamidinium tin triiodide (FASnI
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
806048
Formamidinium iodide
CH5IN2