EMAIL THIS PAGE TO A FRIEND
Journal of microbiology and biotechnology
Nitric Oxide Inhibition and Procollagen Type I Peptide Synthesis Activities of a Phenolic Amide Identified from the Stem of
PMID 28535610
Abstract
The bioactivities of boxthron fruits, a source of oriental medicine, are well known, whereas phytochemical studies of the boxthorn stem are rare. In this study, the stem extract of boxthorn (