EMAIL THIS PAGE TO A FRIEND
Cancer letters
Tg737 regulates epithelial-mesenchymal transition and cancer stem cell properties via a negative feedback circuit between Snail and HNF4α during liver stem cell malignant transformation.
PMID 28536011
Abstract
Determining the origin of liver cancer stem cells is important for treating hepatocellular carcinoma. Tg737 deficiency plays an important role in the malignant transformation of liver stem cells, but the underlying mechanism remains unclear. Here we established a chemical-induced mouse hepatoma model and found that Tg737 and hepatocyte nuclear factor 4-alpha (HNF4α) expression decreased and epithelial-mesenchymal transition (EMT)-related marker expression increased during liver cancer development. To investigate the underlying mechanism, we knocked down Tg737 in WB-F344 (WB) rat hepatic oval cells. Loss of Tg737 resulted in nuclear β-catenin accumulation and activation of the Wnt/β-catenin pathway, which further promoted EMT and the malignant phenotype. XAV939, a β-catenin inhibitor, attenuated WB cell malignant transformation due to Tg737 knockdown. To clarify the relationships of Tg737, the β-catenin pathway, and HNF4α, we inhibited Snail and overexpressed HNF4α after Tg737 knockdown in WB cells and found that Snail and HNF4α comprise a negative feedback circuit. Taken together, the results showed that Tg737 regulates a Wnt/β-catenin/Snail-HNF4α negative feedback circuit, thereby blocking EMT and the malignant transformation of liver stem cells to liver cancer stem cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU064831 MISSION® esiRNA, esiRNA human IFT88 (esiRNA1)