EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Polycyclic aromatic hydrocarbons in urban soil of Novi Sad, Serbia: occurrence and cancer risk assessment.
PMID 28537034
Abstract
Contents of 16 polycyclic aromatic hydrocarbons were analyzed in 30 soil samples from 15 locations in Novi Sad, Serbia, assessing for the first time the corresponding health risks in the Serbian urban zone. Total concentrations were in the range of 22-2247 μg kg
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T82007
1,3,5-Triphenylbenzene, 97%
C24H18