EMAIL THIS PAGE TO A FRIEND
Nature communications
Antigen-specific CD8
PMID 28537251
Abstract
The connection between innate and adaptive immunity is best exemplified by antigen presentation. Although antigen-presenting cells (APCs) are required for antigen receptor-mediated T-cell activation, how T-cells feedback to APCs to sustain an antigen-specific immune response is not completely clear. Here we show that CD8