EMAIL THIS PAGE TO A FRIEND
International journal of cardiology
Prediabetes is not a risk factor for subclinical coronary atherosclerosis.
PMID 28539206
Abstract
There are limited data regarding the influence of glycemic status on the risk of subclinical coronary atherosclerosis on coronary computed tomographic angiography (CCTA) in asymptomatic individuals. We analyzed 6434 asymptomatic individuals who underwent CCTA. The degree and extent of subclinical coronary atherosclerosis were assessed by CCTA, and ≥50% diameter stenosis was defined as significant. Of study participants, 2197 (34.1%), 3122 (48.5%), and 1115 (17.3%) were categorized as normal, prediabetic and diabetic individuals, respectively. Compared with normal individuals, there were no statistically differences in the adjusted odds ratios of prediabetic individuals for significant coronary artery stenosis (0.98, 95% confidence interval [CI] 0.80-1.22, p=0.888), any plaque (0.96, 95% CI 0.86-1.07, p=0.483), calcified plaque (0.90, 95% CI 0.79-1.01, p=0.080), non-calcified plaque (1.02, 95% CI 0.88-1.17, p=0.803), and mixed plaque (1.00, 95% CI 0.82-1.22, p=0.983). However, adjusted odds ratios for significant coronary artery stenosis (1.71, 95% CI 1.34-2.19, p<0.001), any plaque (1.45, 95% CI 1.26-1.68, p<0.001), calcified plaque (1.35, 95% CI 1.15-1.57, p<0.001), non-calcified plaque (1.33, 95% CI 1.11-1.59, p=0.002), and mixed plaque (1.64, 95% CI 1.30-2.07, p<0.001) of diabetic individuals were significantly higher than those of the normal individuals. In asymptomatic individuals, diabetic individuals had a higher risk for subclinical coronary atherosclerosis, but prediabetic individuals were not associated with an increased risk of subclinical coronary atherosclerosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50787
Bisoprolol, analytical standard
C18H31NO4