EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Implications for breast cancer treatment from increased autotaxin production in adipose tissue after radiotherapy.
PMID 28539367
Abstract
We have previously established that adipose tissue adjacent to breast tumors becomes inflamed by tumor-derived cytokines. This stimulates autotaxin (ATX) secretion from adipocytes, whereas breast cancer cells produce insignificant ATX. Lysophosphatidate produced by ATX promotes inflammatory cytokine secretion in a vicious inflammatory cycle, which increases tumor growth and metastasis and decreases response to chemotherapy. We hypothesized that damage to adipose tissue during radiotherapy for breast cancer should promote lysophosphatidic acid (LPA) signaling and further inflammatory signaling, which could potentially protect cancer cells from subsequent fractions of radiation therapy. To test this hypothesis, we exposed rat and human adipose tissue to radiation doses (0.25-5 Gy) that were expected during radiotherapy. This exposure increased mRNA levels for ATX, cyclooxygenase-2, IL-1β, IL-6, IL-10, TNF-α, and LPA
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9867
CGK733, ≥98% (HPLC), solid
C23H18Cl3FN4O3S