EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur physikalische Chemie (Frankfurt am Main, Germany)
Triarylmethyl Radicals: EPR Study of
PMID 28539703
Abstract
Triarylmethyl (TAM) radicals are widely used in Electron Paramagnetic Resonance (EPR) spectroscopy as spin labels and in EPR imaging as spin probes for