EMAIL THIS PAGE TO A FRIEND
International journal of medical sciences
Stabilization of 4E-BP1 by PI3K kinase and its involvement in CHK2 phosphorylation in the cellular response to radiation.
PMID 28539821
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU150481 MISSION® esiRNA, esiRNA human EIF4EBP1 (esiRNA1)