EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
The SapA Protein Is Involved in Resistance to Antimicrobial Peptide PR-39 and Virulence of
PMID 28539918
Abstract
Antimicrobial peptides are essential to the innate immune defense of the mammal against bacterial infection. However, pathogenic bacteria have evolved multiple strategies to resist and evade antimicrobial peptides, which is vital to bacterial survival and colonization in hosts. PR-39 is a linear porcine antimicrobial peptide containing 39 amino acid residues with a high proline content. Resistance to antimicrobial peptide PR-39 has been observed in
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D23602
1,3-Diaminopropane, ≥99%
C3H10N2