EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
A dual-signal sensing system based on organic dyes-LDHs film for fluorescence detection of cysteine.
PMID 28540969
Abstract
It is of great significance to sense cysteine (Cys) in a simple and fast way because of its potential applications in biological processes. In this contribution, we proposed a Cu