EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Modulation of Wetting Gradients by Tuning the Interplay between Surface Structuration and Anisotropic Molecular Layers with Bipolar Electrochemistry.
PMID 28544447
Abstract
A new simple and versatile method for the preparation of surface-wetting gradients is proposed. It is based on the combination of electrode surface structuration introduced by a sacrificial template approach and the formation of a tunable molecular gradient by bipolar electrochemistry. The gradient involves the formation of a self-assembled monolayer on a gold surface by selecting an appropriate thiol molecule and subsequent reductive desorption by means of bipolar electrochemistry. Under these conditions, completion of the reductive desorption process evolves along the bipolar surface with a maximum strength localized at the cathodic edge and a decreasing driving force towards the middle of the surface. The remaining quantity of surface-immobilized thiol, therefore, varies as a function of the axial position, resulting in the formation of a molecular gradient. The surface of the bipolar electrode is characterized at each step of the modification by recording heterogeneous electron transfer. Also, the evolution of static contact angles measured with a water droplet deposited on the surface directly reveals the presence of the wetting gradient, which can be modulated by changing the properties of the thiol. This is exemplified with a long, hydrophobic alkane-thiol and a short, hydrophilic mercaptan.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471364
1-Dodecanethiol, ≥98%
C12H26S