EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Cytotoxicity of the methanol extracts of Elephantopus mollis, Kalanchoe crenata and 4 other Cameroonian medicinal plants towards human carcinoma cells.
PMID 28545532
Abstract
Cancer still constitutes one of the major health concerns globally, causing serious threats on patients, their families, and the healthcare system. In this study, the cytotoxicity of the methanol extract of Elephantopus mollis whole plant (EMW), Enantia chlorantha bark (ECB), Kalanchoe crenata leaves (KCL), Lophira alata bark (LAB), Millettia macrophylla leaves (MML) and Phragmanthera capitata leaves (PCL) towards five human solid cancer cell lines and normal CRL2120 fibroblasts, was evaluated. Extracts were subjected to qualitative chemical screening of their secondary metabolite contents using standard methods. The cytotoxicity of samples was evaluated using neutral red uptake (NR) assay meanwhile caspase activation was detected by caspase-Glo assay. Flow cytometry was used to analyze the cell cycle distribution and the mitochondrial membrane potential (MMP) whilst spectrophotometry was used to measure the levels of reactive oxygen species (ROS). Phytochemical analysis revealed the presence of polyphenols, triterpenes and sterols in all extracts. The IC This study provides evidences of the cytotoxicity of the tested plant extract and highlights the good activity of Elephantopus mollis and Kalanchoe crenata. They deserve more exploration to develop novel cytotoxic drugs.