EMAIL THIS PAGE TO A FRIEND
Biology open
Intra-generational protein malnutrition impairs temporal astrogenesis in rat brain.
PMID 28546341
Abstract
The lack of information on astrogenesis following stressor effect, notwithstanding the imperative roles of astroglia in normal physiology and pathophysiology, incited us to assess temporal astrogenesis and astrocyte density in an intra-generational protein malnutrition (PMN) rat model. Standard immunohistochemical procedures for glial lineage markers and their intensity measurements, and qRT-PCR studies, were performed to reveal the spatio-temporal origin and density of astrocytes. Reduced A