EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Deposition of a Sorbent into a Recession on a Solid Support To Provide a New, Mechanically Robust Solid-Phase Microextraction Device.
PMID 28548491
Abstract
To date, solid-phase microextraction (SPME) fibers used for in vivo bioanalysis can be too fragile and flexible, which limits suitability for direct tissue sampling. As a result, these devices often require a sheathing needle to prepuncture robust sample matrixes and protect the extraction phase from mechanical damage. To address this limitation, a new SPME device is herein presented which incorporates an extraction phase recessed into the body of a solid needle. This device requires no additional support or shielding during puncture events through protective tissue. The presented device was thoroughly tested, being fired at 90 m·s