EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Development of a Sustained-Release Voriconazole-Containing Thermogel for Subconjunctival Injection in Horses.
PMID 28549089
Abstract
To determine in vitro release profiles, transcorneal permeation, and ocular injection characteristics of a voriconazole-containing thermogel suitable for injection into the subconjunctival space (SCS). In vitro release rate of voriconazole (0.3% and 1.5%) from poly (DL-lactide-co-glycolide-b-ethylene glycol-b-DL-lactide-co-glycolide) (PLGA-PEG-PLGA) thermogel was determined for 28 days. A Franz cell diffusion chamber was used to evaluate equine transcorneal and transscleral permeation of voriconazole (1.5% topical solution, 0.3% and 1.5% voriconazole-thermogel) for 24 hours. Antifungal activity of voriconazole released from the 1.5% voriconazole-thermogel was determined via the agar disk diffusion method. Ex vivo equine eyes were injected with liquid voriconazole-thermogel (4°C). Distension of the SCS was assessed ultrasonographically and macroscopically. SCS voriconazole-thermogel injections were performed in a horse 1 week and 2 hours before euthanasia and histopathologic analysis of ocular tissues performed. Voriconazole was released from the PLGA-PEG-PLGA thermogel for more than 21 days in all groups. Release followed first-order kinetics. Voriconazole diffused through the cornea and sclera in all groups. Permeation was greater through the sclerae than corneas. Voriconazole released from the 1.5% voriconazole-thermogel showed antifungal activity in vitro. Voriconazole-thermogel was easily able to be injected into the dorsal SCS where it formed a discrete gel deposit. Voriconazole-thermogel was easily injected in vivo and did not induce any adverse reactions. Voriconazole-containing thermogels have potential application in treatment of keratomycosis. Further research is required to evaluate their performance in vivo.