EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Direct residue analysis of systemic insecticides and some of their relevant metabolites in wines by liquid chromatography - mass spectrometry.
PMID 28549715
Abstract
A direct large volume injection (DI-LVI) high performance liquid chromatography - tandem mass spectrometry (HPLC-MS/MS) method was developed and validated for the quantitative determination of 16 systemic insecticides and their main plant metabolites. The assays were conducted on commercial red and white wines made from grapes grown in major wine-producing regions nationally and internationally. Using a 1:20 dilution and an injection volume of 800μL, a limit of quantitation (LOQ) of 1μgL