EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
HIV-1 decreases Nrf2/ARE activity and phagocytic function in alveolar macrophages.
PMID 28550120
Abstract
Respiratory complications occur frequently in individuals living with human immunodeficiency-1 virus (HIV) infection, and there is evidence that HIV-related oxidative stress impairs alveolar macrophage immune function. We hypothesized that nuclear factor (erythroid-derived 2)-like 2 (Nrf2), a master transcription factor that activates the antioxidant response element (ARE) and regulates antioxidant defenses, has an important role in alveolar macrophage (AMs) immune dysfunction in individuals with HIV infections. To test that hypothesis, we analyzed human monocyte-derived macrophages (MDMs) that were either infected with HIV-1 or were exposed to the HIV-related proteins gp120 and Tat ex vivo and determined that either stress affected the expression of Nrf2 and the Nrf2-ARE-dependent genes for NAD(P)H dehydrogenase, quinone 1 (