EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Ex-vivo measurement of scalp follicular infundibulum delivery of zinc pyrithione and climbazole from an anti-dandruff shampoo.
PMID 28550716
Abstract
Efficient delivery of anti-dandruff (AD) actives into the scalp follicular infundibulum as well as onto the scalp surface is critical for the efficacy of AD shampoos. A method involving scalp cyanoacrylate (CA) biopsy sampling, a tailor made cutting device, ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) analysis, scanning electron microscopy (SEM) measurement and Raman imaging has been developed for the measurement of delivery of zinc pyrithione (ZPT) and climbazole (CBZ) from an AD shampoo into the scalp follicular infundibulum. Scalp CA biopsy enables the sampling of ZPT and CBZ delivered into the scalp follicular infundibula as well as onto the scalp surface. Raman imaging of scalp CA biopsy samples allows the visualization of the spatial distribution of ZPT and CBZ deposited on the scalp. A tailor made cutting device enables the separation of the scalp follicular infundibulum sample (20μm below the scalp surface) from the scalp surface samples (including top 20μm of infundibula). UHPLC-MS/MS was used as a sensitive and specific methodology enabling the quantification of ZPT and CBZ without interference. Using this method, both ZPT and CBZ were successfully quantified and spacially visualized within the scalp follicular infundibulum, after scalp was washed with an AD shampoo.