EMAIL THIS PAGE TO A FRIEND
Food chemistry
Enzyme-assisted extraction enhancing the umami taste amino acids recovery from several cultivated mushrooms.
PMID 28551231
Abstract
In this study, enzyme-assisted extraction was performed to extract umami taste and total free amino acids (FAAs) from the six different mushrooms including shiitake (Lentinus edodes), oyster (Pleurotus ostreatus), tea tree (Agrocybe aegerita) and, white, brown and portobello champignons (Agaricus bisporus). β-Glucanase and Flavourzyme® were used as the enzymes for cell wall and proteins hydrolysis, respectively. It was found that β-glucanase treatment alone did not enhance the extraction efficiency, however in combination, β-glucanase and Flavourzyme® enhanced the extraction efficiency significantly up to 20-fold compared to conventional HCl mediated extraction, depending on the mushroom species. The optimal conditions for the enzyme treatment were: water as extraction solvent (initial pH = 7), enzyme concentration of 5% v/w each of β-glucanase and Flavourzyme®, temperature 50°C and an incubation time of 1h. White and brown champignons were found to be the richest source of umami taste FAAs (26.75±1.07 and 25.6±0.9mg/g DM, respectively).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I8754
L-allo-Isoleucine
C6H13NO2