EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Butylated hydroxyanisole alters rat 5α-reductase and 3α-hydroxysteroid dehydrogenase: Implications for influences of neurosteroidogenesis.
PMID 28552457
Abstract
Butylated hydroxyanisole is a synthetic antioxidant. It may affect the function of the nerve system. The objective of the present study is to investigate the direct effects of butylated hydroxyanisole on rat brain neurosteroidogenic 5α-reductase 1 (SRD5A1), 3α-hydroxysteroid dehydrogenase (AKR1C14), and retinol dehydrogenase 2 (RDH2). Rat SRD5A1, AKR1C14, and RDH2 were cloned and expressed in COS1 cells, and the effects of butylated hydroxyanisole on these enzyme activities were measured. Butylated hydroxyanisole inhibited SRD5A1, AKR1C14, and RDH2 with IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1253
Butylated hydroxyanisole, ≥98.5%
C11H16O2