EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Both non-covalent and covalent interactions were involved in the mechanism of detoxifying effects of persimmon tannin on Chinese cobra PLA
PMID 28552595
Abstract
Persimmon tannin (PT) has been shown to inhibit snake venom activities and toxicities both in vitro and in vivo. To clarify the detoxifying mechanism of PT on snake venom, the interaction of characteristic structural elements of PT (EGCG, ECG, EGCG dimer and ECG dimer) and Chinese cobra phospholipase A